Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCAAGGCCTCCAAGATGAGCTACAC[A/G]TTGGACTCGCTGGGCAACCCGTCCG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 162250 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
NEFM PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
NEFM - neurofilament, medium polypeptide | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001105541.1 | 45 | Intron | NP_001099011.1 | |||
NM_005382.2 | 45 | Silent Mutation | ACA,ACG | T,T 4 | NP_005373.2 |