Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TCATACCACACTCCTCACACTCATA[A/G]GGCTTCTCTCCAGTGTGGACTCGCT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 601069 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
ZNF239 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
ZNF239 - zinc finger protein 239 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001099282.1 | 1125 | Silent Mutation | CCC,CCT | P,P 318 | NP_001092752.1 | |
NM_001099283.1 | 1125 | Silent Mutation | CCC,CCT | P,P 318 | NP_001092753.1 | |
NM_001099284.1 | 1125 | Silent Mutation | CCC,CCT | P,P 318 | NP_001092754.1 | |
NM_001324347.1 | 1125 | Silent Mutation | CCC,CCT | P,P 318 | NP_001311276.1 | |
NM_001324348.1 | 1125 | Silent Mutation | CCC,CCT | P,P 318 | NP_001311277.1 | |
NM_001324349.1 | 1125 | Silent Mutation | CCC,CCT | P,P 318 | NP_001311278.1 | |
NM_001324350.1 | 1125 | Silent Mutation | CCC,CCT | P,P 318 | NP_001311279.1 | |
NM_001324351.1 | 1125 | Silent Mutation | CCC,CCT | P,P 318 | NP_001311280.1 | |
NM_001324352.1 | 1125 | Silent Mutation | CCC,CCT | P,P 360 | NP_001311281.1 | |
NM_001324353.1 | 1125 | Silent Mutation | CCC,CCT | P,P 431 | NP_001311282.1 | |
NM_005674.2 | 1125 | Silent Mutation | CCC,CCT | P,P 318 | NP_005665.2 | |
XM_005271832.2 | 1125 | Silent Mutation | CCC,CCT | P,P 318 | XP_005271889.1 | |
XM_006718001.2 | 1125 | Silent Mutation | CCC,CCT | P,P 360 | XP_006718064.1 | |
XM_006718003.3 | 1125 | Silent Mutation | CCC,CCT | P,P 318 | XP_006718066.1 | |
XM_011540232.2 | 1125 | Silent Mutation | CCC,CCT | P,P 462 | XP_011538534.1 | |
XM_011540234.2 | 1125 | Silent Mutation | CCC,CCT | P,P 360 | XP_011538536.1 | |
XM_011540235.2 | 1125 | Silent Mutation | CCC,CCT | P,P 360 | XP_011538537.1 | |
XM_011540236.2 | 1125 | Silent Mutation | CCC,CCT | P,P 360 | XP_011538538.1 | |
XM_011540237.2 | 1125 | Silent Mutation | CCC,CCT | P,P 360 | XP_011538539.1 | |
XM_011540238.2 | 1125 | Silent Mutation | CCC,CCT | P,P 318 | XP_011538540.1 | |
XM_017016740.1 | 1125 | Silent Mutation | CCC,CCT | P,P 360 | XP_016872229.1 |