Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TAGTTTTCAAGCTTGTGTATGTTAC[C/T]GTGTTTCTCACATCACTCTCATAAC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 612123 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
PNPLA8 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
PNPLA8 - patatin like phospholipase domain containing 8 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001256007.2 | 3258 | Silent Mutation | ACA,ACG | T,T 670 | NP_001242936.1 | |
NM_001256008.2 | 3258 | Silent Mutation | ACA,ACG | T,T 670 | NP_001242937.1 | |
NM_001256009.2 | 3258 | Silent Mutation | ACA,ACG | T,T 608 | NP_001242938.1 | |
NM_001256010.2 | 3258 | Silent Mutation | ACA,ACG | T,T 570 | NP_001242939.1 | |
NM_001256011.2 | 3258 | Silent Mutation | ACA,ACG | T,T 570 | NP_001242940.1 | |
NM_015723.4 | 3258 | Silent Mutation | ACA,ACG | T,T 670 | NP_056538.1 | |
XM_005250396.4 | 3258 | Silent Mutation | ACA,ACG | T,T 608 | XP_005250453.1 | |
XM_011516274.2 | 3258 | Silent Mutation | ACA,ACG | T,T 698 | XP_011514576.1 | |
XM_011516275.2 | 3258 | Silent Mutation | ACA,ACG | T,T 636 | XP_011514577.1 | |
XM_017012261.1 | 3258 | Silent Mutation | ACA,ACG | T,T 508 | XP_016867750.1 |