Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGGATTATGCTCGCCCCACAACACT[A/G]GGGTGTGGCAGTGAGAAGAAATCTC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 606872 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
CBLL1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
CBLL1 - Cbl proto-oncogene like 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001284291.1 | 119 | Intron | NP_001271220.1 | |||
NM_024814.3 | 119 | Intron | NP_079090.2 | |||
XM_011516580.2 | 119 | Intron | XP_011514882.1 | |||
XM_017012643.1 | 119 | UTR 5 | XP_016868132.1 | |||
XM_017012644.1 | 119 | UTR 5 | XP_016868133.1 | |||
XM_017012645.1 | 119 | Intron | XP_016868134.1 |