Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCGGGACGGGAGGGTGGAGCCAAGT[A/C]CCCCCGCGGCCGGTTCGGGAGCTTT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||
Literature Links: |
LOC105376204 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
LOC105376204 - uncharacterized LOC105376204 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
XM_017015409.1 | 15 | Missense Mutation | XP_016870898.1 |
ZNF462 - zinc finger protein 462 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_021224.4 | 15 | Intron | NP_067047.4 | |||
XM_006717209.3 | 15 | Intron | XP_006717272.1 | |||
XM_006717211.3 | 15 | Intron | XP_006717274.1 | |||
XM_006717212.3 | 15 | UTR 5 | XP_006717275.1 | |||
XM_006717215.3 | 15 | Intron | XP_006717278.1 | |||
XM_006717216.3 | 15 | Intron | XP_006717279.1 | |||
XM_006717218.3 | 15 | Intron | XP_006717281.1 | |||
XM_011518892.2 | 15 | Intron | XP_011517194.1 | |||
XM_017014996.1 | 15 | Intron | XP_016870485.1 | |||
XM_017014997.1 | 15 | Intron | XP_016870486.1 | |||
XM_017014998.1 | 15 | Intron | XP_016870487.1 | |||
XM_017014999.1 | 15 | Intron | XP_016870488.1 |