Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GTCCAGACCTGACAGAAGGGAAGAG[G/A]CCAGCATTGGGATGAAATCCAGTGA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
ARMCX5 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese)
|
||||||
EUR
|
||||||||
AMR
|
ARMCX5 - armadillo repeat containing, X-linked 5 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001168478.1 | 1124 | Missense Mutation | ACC,GCC | T,A 156 | NP_001161950.1 | |
NM_001168479.1 | 1124 | Missense Mutation | ACC,GCC | T,A 156 | NP_001161951.1 | |
NM_001168480.1 | 1124 | Missense Mutation | ACC,GCC | T,A 156 | NP_001161952.1 | |
NM_001168482.1 | 1124 | Missense Mutation | ACC,GCC | T,A 156 | NP_001161954.1 | |
NM_001168485.1 | 1124 | Missense Mutation | ACC,GCC | T,A 156 | NP_001161957.1 | |
NM_022838.3 | 1124 | Missense Mutation | ACC,GCC | T,A 156 | NP_073749.2 |
ARMCX5-GPRASP2 - ARMCX5-GPRASP2 readthrough | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001199818.1 | 1124 | Intron | NP_001186747.1 |
Set Membership: |
HapMap |