Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GTCATCAGTAAAGCCAGAACAAGGG[A/G]AACAAATTTATCATAAGCAGCATCT
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 603924 MIM: 602873 | |||||||||||||||||||||||||||||||||||||||||
Literature Links: |
HABP2 PubMed Links | |||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
HABP2 - hyaluronan binding protein 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001177660.2 | Intron | NP_001171131.1 | ||||
NM_004132.4 | Intron | NP_004123.1 |
NRAP - nebulin related anchoring protein | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001261463.1 | Intron | NP_001248392.1 | ||||
NM_001322945.1 | Intron | NP_001309874.1 | ||||
NM_006175.4 | Intron | NP_006166.3 | ||||
NM_198060.3 | Intron | NP_932326.2 | ||||
XM_005269864.2 | Intron | XP_005269921.1 | ||||
XM_005269865.2 | Intron | XP_005269922.1 | ||||
XM_005269867.2 | Intron | XP_005269924.1 | ||||
XM_006717870.2 | Intron | XP_006717933.1 | ||||
XM_011539832.2 | Intron | XP_011538134.1 |
Set Membership: |
HapMap |