Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 601964 MIM: 604497 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
DNAJC7 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||||||||
---|---|---|---|---|---|---|---|---|---|---|---|
Global
|
Caucasian
|
CEPH (CEU)
|
|||||||||
EAS
|
African American
|
YRI (Yoruba)
|
|||||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||||||||
EUR
|
|||||||||||
AMR
|
DNAJC7 - DnaJ heat shock protein family (Hsp40) member C7 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001144766.2 | Intron | NP_001138238.1 | ||||
NM_003315.3 | Intron | NP_003306.3 | ||||
XM_011525167.2 | Intron | XP_011523469.1 | ||||
XM_011525168.2 | Intron | XP_011523470.1 | ||||
XM_011525169.2 | Intron | XP_011523471.1 | ||||
XM_017024994.1 | Intron | XP_016880483.1 |
NKIRAS2 - NFKB inhibitor interacting Ras like 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001001349.2 | Intron | NP_001001349.1 | ||||
NM_001144927.1 | Intron | NP_001138399.1 | ||||
NM_001144928.1 | Intron | NP_001138400.1 | ||||
NM_001144929.1 | Intron | NP_001138401.1 | ||||
NM_017595.5 | Intron | NP_060065.2 | ||||
XM_005257251.1 | Intron | XP_005257308.1 |
ZNF385C - zinc finger protein 385C | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
Set Membership: |
HapMap Validated |