Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGCGGACAGATGTCCCTGAAGGCCC[A/G]AGGGACACCAGCCGCTATGCCAGGC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 608568 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
MYH14 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
MYH14 - myosin, heavy chain 14, non-muscle | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001077186.1 | 22 | Intron | NP_001070654.1 | |||
NM_001145809.1 | 22 | Intron | NP_001139281.1 | |||
NM_024729.3 | 22 | Intron | NP_079005.3 | |||
XM_006723386.3 | 22 | Intron | XP_006723449.1 | |||
XM_011527320.2 | 22 | Missense Mutation | CAA,CGA | Q,R 6 | XP_011525622.1 | |
XM_011527321.2 | 22 | Missense Mutation | CAA,CGA | Q,R 6 | XP_011525623.1 | |
XM_011527323.2 | 22 | Intron | XP_011525625.1 |