Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGGATATTCAATGAGCATTATTTTT[C/T]CACAGCTTGGAACATCACCCCTCCA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
19 submissions
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||
Literature Links: |
GABPB2 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
GABPB2 - GA binding protein transcription factor beta subunit 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001323906.1 | Intron | NP_001310835.1 | ||||
NM_001323907.1 | Intron | NP_001310836.1 | ||||
NM_001323908.1 | Intron | NP_001310837.1 | ||||
NM_001323909.1 | Intron | NP_001310838.1 | ||||
NM_001323910.1 | Intron | NP_001310839.1 | ||||
NM_001323911.1 | Intron | NP_001310840.1 | ||||
NM_001323912.1 | Intron | NP_001310841.1 | ||||
NM_001323913.1 | Intron | NP_001310842.1 | ||||
NM_144618.2 | Intron | NP_653219.1 | ||||
XM_017000246.1 | Intron | XP_016855735.1 | ||||
XM_017000247.1 | Intron | XP_016855736.1 | ||||
XM_017000248.1 | Intron | XP_016855737.1 | ||||
XM_017000249.1 | Intron | XP_016855738.1 | ||||
XM_017000250.1 | Intron | XP_016855739.1 | ||||
XM_017000251.1 | Intron | XP_016855740.1 | ||||
XM_017000252.1 | Intron | XP_016855741.1 | ||||
XM_017000253.1 | Intron | XP_016855742.1 | ||||
XM_017000254.1 | Intron | XP_016855743.1 |