Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AACTGAAGAACATATATTTATACCC[C/T]AAAGAAAATTATACAAAGTTATTTC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 607520 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
FRYL PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
FRYL - FRY like transcription coactivator | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_015030.1 | Intron | NP_055845.1 | ||||
XM_005248090.4 | Intron | XP_005248147.1 | ||||
XM_005248093.4 | Intron | XP_005248150.1 | ||||
XM_011513680.2 | Intron | XP_011511982.1 | ||||
XM_011513681.2 | Intron | XP_011511983.1 | ||||
XM_011513682.2 | Intron | XP_011511984.1 | ||||
XM_011513683.2 | Intron | XP_011511985.1 | ||||
XM_011513684.2 | Intron | XP_011511986.1 | ||||
XM_011513685.2 | Intron | XP_011511987.1 | ||||
XM_017008046.1 | Intron | XP_016863535.1 |
ZAR1 - zygote arrest 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |