Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CAGCACCTCTCTTCTCTCACCAGAA[A/C]CTTGTGTCCAAGGAGGTCCTTCAGG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
30 submissions
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||
Literature Links: |
DCST1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
DCST1 - DC-STAMP domain containing 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001143687.2 | 52 | UTR 5 | NP_001137159.1 | |||
NM_152494.3 | 52 | UTR 5 | NP_689707.2 |
DCST2 - DC-STAMP domain containing 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_144622.2 | 52 | Intron | NP_653223.2 | |||
XM_011509188.2 | 52 | Intron | XP_011507490.1 | |||
XM_011509189.2 | 52 | Intron | XP_011507491.1 |