Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AAGTCATCTAATCTAATAATAGAGA[C/T]ATTGCCCAGACATTCTGTACACACC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 613081 MIM: 187680 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
KDM1B PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
KDM1B - lysine demethylase 1B | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_153042.3 | Intron | NP_694587.3 | ||||
XM_005248925.1 | Intron | XP_005248982.1 | ||||
XM_005248926.1 | Intron | XP_005248983.1 | ||||
XM_005248928.1 | Intron | XP_005248985.1 | ||||
XM_011514385.1 | Intron | XP_011512687.1 | ||||
XM_011514386.1 | Intron | XP_011512688.1 | ||||
XM_017010440.1 | Intron | XP_016865929.1 | ||||
XM_017010441.1 | Intron | XP_016865930.1 | ||||
XM_017010442.1 | Intron | XP_016865931.1 | ||||
XM_017010443.1 | Intron | XP_016865932.1 | ||||
XM_017010445.1 | Intron | XP_016865934.1 | ||||
XM_017010446.1 | Intron | XP_016865935.1 |
TPMT - thiopurine S-methyltransferase | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
Set Membership: |
HapMap |