Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCGAGGGAGATGGGAAACGGGGCGG[A/G]TGGCGCCCAGCCCAGCCCTAACTGC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 604237 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
C19orf60 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
C19orf60 - chromosome 19 open reading frame 60 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001100418.1 | 691 | UTR 3 | NP_001093888.1 | |||
NM_001100419.1 | 691 | UTR 3 | NP_001093889.1 | |||
XM_005259977.1 | 691 | UTR 3 | XP_005260034.1 |
CRLF1 - cytokine receptor like factor 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_004750.4 | 691 | Intron | NP_004741.1 | |||
XM_011528422.2 | 691 | Intron | XP_011526724.1 | |||
XM_011528423.2 | 691 | Intron | XP_011526725.1 | |||
XM_011528424.2 | 691 | Intron | XP_011526726.1 |