Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CACCTCCTTCCCACCCCCTCAGCCT[C/T]GGTCCTCTGTGGAGAGGCTGTGTCG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 167405 | ||||||||||||||||||||
Literature Links: |
LOC100507472 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
LOC100507472 - uncharacterized LOC100507472 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
PCSK6 - proprotein convertase subtilisin/kexin type 6 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001291309.1 | Intron | NP_001278238.1 | ||||
NM_002570.4 | Intron | NP_002561.1 | ||||
NM_138319.3 | Intron | NP_612192.1 | ||||
NM_138322.3 | Intron | NP_612195.1 | ||||
NM_138323.2 | Intron | NP_612196.1 | ||||
NM_138324.2 | Intron | NP_612197.1 | ||||
NM_138325.3 | Intron | NP_612198.2 |