Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 607993 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
SUGP2 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
SUGP2 - SURP and G-patch domain containing 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001017392.4 | Intron | NP_001017392.2 | ||||
NM_001321697.1 | Intron | NP_001308626.1 | ||||
NM_001321698.1 | Intron | NP_001308627.1 | ||||
NM_001321699.1 | Intron | NP_001308628.1 | ||||
NM_014884.4 | Intron | NP_055699.2 | ||||
XM_017026137.1 | Intron | XP_016881626.1 |
Set Membership: |
HapMap |