Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ACTTCGAATAAAGTTGTTCCAATGA[A/G]GACTGCATAGTTTCCAAATAACCAG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 604983 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
MILR1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
MILR1 - mast cell immunoglobulin like receptor 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001085423.1 | 1367 | Intron | NP_001078892.1 | |||
NM_001291316.1 | 1367 | Intron | NP_001278245.1 | |||
NM_001291317.1 | 1367 | Intron | NP_001278246.1 |
POLG2 - DNA polymerase gamma 2, accessory subunit | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_007215.3 | 1367 | Silent Mutation | TCC,TCT | S,S 423 | NP_009146.2 | |
XM_006721651.2 | 1367 | Intron | XP_006721714.1 | |||
XM_017024072.1 | 1367 | Intron | XP_016879561.1 |