Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TCTTGCTGGGTGAGATGGCAGTGTG[C/T]GCAAAAAAGCGCCCCCCAGGTAAGA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
18 submissions
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 606578 MIM: 605867 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
AQP10 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
AQP10 - aquaporin 10 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
ATP8B2 - ATPase phospholipid transporting 8B2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001005855.1 | 211 | Silent Mutation | TGC,TGT | C,C 4 | NP_001005855.1 | |
NM_020452.3 | 211 | Silent Mutation | TGC,TGT | C,C 37 | NP_065185.1 | |
XM_005245355.2 | 211 | Intron | XP_005245412.1 | |||
XM_005245356.3 | 211 | Silent Mutation | TGC,TGT | C,C 4 | XP_005245413.1 | |
XM_017001871.1 | 211 | Intron | XP_016857360.1 | |||
XM_017001872.1 | 211 | Silent Mutation | TGC,TGT | C,C 4 | XP_016857361.1 |