Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GCCCCGGGCTTTCTCCACTCGGATC[T/C]CGCCCCGTTTGGGTCCCGGGGAGGG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||
Literature Links: |
ZNF747 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
ZNF747 - zinc finger protein 747 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001305018.1 | 373 | Missense Mutation | AGA,GGA | R,G 28 | NP_001291947.1 | |
NM_001305019.1 | 373 | Missense Mutation | AGA,GGA | R,G 28 | NP_001291948.1 | |
NM_001305020.1 | 373 | Missense Mutation | AGA,GGA | R,G 28 | NP_001291949.1 | |
NM_023931.3 | 373 | Missense Mutation | AGA,GGA | R,G 28 | NP_076420.1 |
ZNF768 - zinc finger protein 768 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |