Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCACTCTTCCGCTCTCGAACCCTCT[A/G]CAGGGGCCTGGACGCTAATCCTCAG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 602128 MIM: 602220 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
GAS2L1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
GAS2L1 - growth arrest specific 2 like 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
RASL10A - RAS like family 10 member A | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_006477.4 | Intron | NP_006468.1 | ||||
XM_011529821.1 | Intron | XP_011528123.1 | ||||
XM_011529822.1 | Intron | XP_011528124.1 | ||||
XM_011529823.1 | Intron | XP_011528125.1 |