Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGGCTTCTCCCCGGTGTGGATCCTC[C/T]GGTGCTGAATCAAGTACGTGCTCTG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 194526 | ||||||||||||||||||||
Literature Links: |
ZNF34 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
ZNF34 - zinc finger protein 34 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001286769.1 | 1676 | Missense Mutation | CAG,CGG | Q,R 501 | NP_001273698.1 | |
NM_001286770.1 | 1676 | Missense Mutation | CAG,CGG | Q,R 482 | NP_001273699.1 | |
NM_030580.4 | 1676 | Missense Mutation | CAG,CGG | Q,R 522 | NP_085057.3 | |
XM_011517314.2 | 1676 | Missense Mutation | CAG,CGG | Q,R 522 | XP_011515616.1 | |
XM_011517315.2 | 1676 | Missense Mutation | CAG,CGG | Q,R 501 | XP_011515617.1 | |
XM_011517316.2 | 1676 | Missense Mutation | CAG,CGG | Q,R 501 | XP_011515618.1 | |
XM_011517318.2 | 1676 | Missense Mutation | CAG,CGG | Q,R 501 | XP_011515620.1 | |
XM_011517320.2 | 1676 | Missense Mutation | CAG,CGG | Q,R 461 | XP_011515622.1 | |
XM_017013872.1 | 1676 | Missense Mutation | CAG,CGG | Q,R 522 | XP_016869361.1 | |
XM_017013873.1 | 1676 | Missense Mutation | CAG,CGG | Q,R 522 | XP_016869362.1 | |
XM_017013874.1 | 1676 | Missense Mutation | CAG,CGG | Q,R 522 | XP_016869363.1 | |
XM_017013875.1 | 1676 | Missense Mutation | CAG,CGG | Q,R 501 | XP_016869364.1 | |
XM_017013876.1 | 1676 | Missense Mutation | CAG,CGG | Q,R 461 | XP_016869365.1 |