Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ACCCCACCCTCCAGGGCAGTGCCAC[C/T]GTCAGCCCCGTGGGGAGGCCGTGCC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 602267 | ||||||||||||||||||||
Literature Links: |
ADAM8 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
ADAM8 - ADAM metallopeptidase domain 8 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001109.4 | Intron | NP_001100.3 | ||||
NM_001164489.1 | Intron | NP_001157961.1 | ||||
NM_001164490.1 | Intron | NP_001157962.1 | ||||
XM_011539117.2 | Intron | XP_011537419.2 | ||||
XM_017015465.1 | Intron | XP_016870954.1 | ||||
XM_017015466.1 | Intron | XP_016870955.1 |