Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCTGGAAAAAGTCCTGGGAAAACCC[A/G]CCAAGCAACAAAGCCCTCTTACCAA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 600294 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
ADCY6 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
ADCY6 - adenylate cyclase 6 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_015270.4 | Intron | NP_056085.1 | ||||
XM_006719210.3 | Intron | XP_006719273.1 | ||||
XM_017018743.1 | Intron | XP_016874232.1 |
LINC00935 - long intergenic non-protein coding RNA 935 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
MIR4701 - microRNA 4701 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |