Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGTTCCCCACGCTCCCTTCCTCCTC[C/T]TCCTCCTCCTCCCCACCGAAGTCAG
Species: |
Human | |||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||
Phenotype: |
MIM: 606458 MIM: 611430 | |||||||||||||||||||||||
Literature Links: |
CSRNP1 PubMed Links | |||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR - Not Available | |||||
AMR - Not Available |
CSRNP1 - cysteine and serine rich nuclear protein 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001320559.1 | 1438 | Silent Mutation | GAA,GAG | E,E 428 | NP_001307488.1 | |
NM_001320560.1 | 1438 | Silent Mutation | GAA,GAG | E,E 408 | NP_001307489.1 | |
NM_033027.3 | 1438 | Silent Mutation | GAA,GAG | E,E 408 | NP_149016.2 | |
XM_017007048.1 | 1438 | Silent Mutation | GAA,GAG | E,E 408 | XP_016862537.1 | |
XM_017007049.1 | 1438 | Silent Mutation | GAA,GAG | E,E 148 | XP_016862538.1 |
MIR6822 - microRNA 6822 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
TTC21A - tetratricopeptide repeat domain 21A | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |