Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 609118 | |||||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
PDCD10 PubMed Links | |||||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||||||||
---|---|---|---|---|---|---|---|---|---|---|---|
Global
|
Caucasian
|
CEPH (CEU) - Not Available | |||||||||
EAS
|
African American
|
YRI (Yoruba)
|
|||||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
|||||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
|||||||||
EUR
|
|||||||||||
AMR
|
PDCD10 - programmed cell death 10 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_007217.3 | Intron | NP_009148.2 | ||||
NM_145859.1 | Intron | NP_665858.1 | ||||
NM_145860.1 | Intron | NP_665859.1 | ||||
XM_005247086.4 | Intron | XP_005247143.1 | ||||
XM_005247087.4 | Intron | XP_005247144.1 | ||||
XM_005247088.3 | Intron | XP_005247145.1 | ||||
XM_006713485.3 | Intron | XP_006713548.1 | ||||
XM_011512368.2 | Intron | XP_011510670.1 | ||||
XM_011512369.2 | Intron | XP_011510671.1 | ||||
XM_017005644.1 | Intron | XP_016861133.1 | ||||
XM_017005645.1 | Intron | XP_016861134.1 |