Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 603461 | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
CDC16 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||||||||
---|---|---|---|---|---|---|---|---|---|---|---|
Global
|
Caucasian
|
CEPH (CEU)
|
|||||||||
EAS
|
African American
|
YRI (Yoruba)
|
|||||||||
SAS
|
Japanese
|
JPT (Japanese)
|
|||||||||
AFR
|
Chinese
|
CHB (Han Chinese)
|
|||||||||
EUR
|
|||||||||||
AMR
|
CDC16 - cell division cycle 16 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
XM_005266206.1 | 515 | Silent Mutation | GAC,GAT | D,D 110 | XP_005266263.1 | |
XM_006719992.1 | 515 | Silent Mutation | GAC,GAT | D,D 109 | XP_006720055.1 | |
XM_006719993.1 | 515 | Silent Mutation | GAC,GAT | D,D 16 | XP_006720056.1 | |
XM_011534851.1 | 515 | Silent Mutation | GAC,GAT | D,D 110 | XP_011533153.1 | |
XM_011534853.1 | 515 | Silent Mutation | GAC,GAT | D,D 110 | XP_011533155.1 | |
XM_017020830.1 | 515 | Silent Mutation | GAC,GAT | D,D 110 | XP_016876319.1 | |
XM_017020831.1 | 515 | Silent Mutation | GAC,GAT | D,D 110 | XP_016876320.1 | |
XM_017020832.1 | 515 | Silent Mutation | GAC,GAT | D,D 16 | XP_016876321.1 | |
XM_017020833.1 | 515 | Silent Mutation | GAC,GAT | D,D 16 | XP_016876322.1 |
MIR548AR - microRNA 548ar | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
Set Membership: |
HapMap Validated |