Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGGGCAGTCTACAGGGGAGGATGCA[C/T]GAGGGGCAGGCAGGCAGGGGGTGCT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 615746 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
CFAP100 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
CFAP100 - cilia and flagella associated protein 100 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
LOC105374089 - uncharacterized LOC105374089 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
ZXDC - ZXD family zinc finger C | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001040653.3 | Intron | NP_001035743.1 | ||||
NM_025112.4 | Intron | NP_079388.3 | ||||
XM_005247757.3 | Intron | XP_005247814.1 | ||||
XM_006713741.2 | Intron | XP_006713804.1 | ||||
XM_011513119.2 | Intron | XP_011511421.1 |