Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TACCCACTCACCTACCACCCGCCCA[C/G]ACGCTCGCGAAGGCCGACGTAAAAG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 613263 MIM: 613117 MIM: 613264 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
SNHG5 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
SNHG5 - small nucleolar RNA host gene 5 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
SNORD50A - small nucleolar RNA, C/D box 50A | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
SNORD50B - small nucleolar RNA, C/D box 50B | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |