Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ACAGGAGCTCTTCCAAGCTGGCACA[A/G]CTGATCTCAGGGTTGGCAGACAGAT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 604546 MIM: 611952 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
MIR6893 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
MIR6893 - microRNA 6893 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
TONSL - tonsoku-like, DNA repair protein | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_013432.4 | 3885 | Silent Mutation | AGC,AGT | S,S 1290 | NP_038460.4 | |
XM_011517048.2 | 3885 | Silent Mutation | AGC,AGT | S,S 966 | XP_011515350.1 | |
XM_011517049.2 | 3885 | Silent Mutation | AGC,AGT | S,S 954 | XP_011515351.1 | |
XM_011517050.2 | 3885 | Intron | XP_011515352.1 |
TONSL-AS1 - TONSL antisense RNA 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
VPS28 - VPS28, ESCRT-I subunit | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |