Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGCAGGACACGCCTTTCCTGACAGG[A/C]CATGGGCGCTTGAGTGAGCAGCAGG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||
Literature Links: |
CARD19 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
CARD19 - caspase recruitment domain family member 19 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001318010.1 | 280 | Silent Mutation | GGA,GGC | G,G 19 | NP_001304939.1 | |
NM_001318011.1 | 280 | Silent Mutation | GGA,GGC | G,G 19 | NP_001304940.1 | |
NM_032310.4 | 280 | Silent Mutation | GGA,GGC | G,G 19 | NP_115686.3 |