Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 610937 | |||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
RPGRIP1L PubMed Links | |||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese)
|
||||||
EUR
|
||||||||
AMR
|
RPGRIP1L - RPGRIP1 like | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
XM_005255867.1 | 3980 | Silent Mutation | GAC,GAT | D,D 1278 | XP_005255924.1 | |
XM_005255868.1 | 3980 | Silent Mutation | GAC,GAT | D,D 1270 | XP_005255925.1 | |
XM_011522970.1 | 3980 | Intron | XP_011521272.1 | |||
XM_011522971.2 | 3980 | Intron | XP_011521273.1 | |||
XM_011522973.2 | 3980 | Intron | XP_011521275.1 | |||
XM_017023094.1 | 3980 | Silent Mutation | GAC,GAT | D,D 1316 | XP_016878583.1 | |
XM_017023095.1 | 3980 | Silent Mutation | GAC,GAT | D,D 1236 | XP_016878584.1 | |
XM_017023096.1 | 3980 | Intron | XP_016878585.1 | |||
XM_017023097.1 | 3980 | Intron | XP_016878586.1 | |||
XM_017023098.1 | 3980 | Silent Mutation | GAC,GAT | D,D 727 | XP_016878587.1 | |
XM_017023099.1 | 3980 | Silent Mutation | GAC,GAT | D,D 727 | XP_016878588.1 | |
XM_017023100.1 | 3980 | Intron | XP_016878589.1 |
Set Membership: |
HapMap |