Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AATCCTAGGCGCGGATCTCGCGTTT[C/G]CGGCCGGAAGCTTCTCCAGCCTTTC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 604904 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
TAF1B PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
TAF1B - TATA-box binding protein associated factor, RNA polymerase I subunit B | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001318976.1 | 12 | UTR 5 | NP_001305905.1 | |||
NM_001318977.1 | 12 | UTR 5 | NP_001305906.1 | |||
NM_005680.2 | 12 | UTR 5 | NP_005671.2 | |||
XM_011510410.2 | 12 | Intron | XP_011508712.1 | |||
XM_011510411.1 | 12 | Intron | XP_011508713.1 |