Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GTTTAAAATTGAGAAGAATAGAGTG[C/T]TAACTGGACATGGGATTTGAAATTC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||
Literature Links: |
CEP95 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
CEP95 - centrosomal protein 95 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001316990.1 | Intron | NP_001303919.1 | ||||
NM_138363.2 | Intron | NP_612372.1 | ||||
XM_005257779.3 | Intron | XP_005257836.1 | ||||
XM_005257780.2 | Intron | XP_005257837.1 | ||||
XM_006722166.3 | Intron | XP_006722229.1 | ||||
XM_011525458.2 | Intron | XP_011523760.1 | ||||
XM_017025287.1 | Intron | XP_016880776.1 | ||||
XM_017025288.1 | Intron | XP_016880777.1 |