Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGCAAGGATATTTAATTTACTTCCA[C/T]ATTTTTTTATTAGTAAAATGCATTT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 603760 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
HUS1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
HUS1 - HUS1 checkpoint clamp component | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
SUN3 - Sad1 and UNC84 domain containing 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001030019.1 | 1328 | UTR 3 | NP_001025190.1 | |||
NM_001284350.1 | 1328 | UTR 3 | NP_001271279.1 | |||
NM_152782.3 | 1328 | UTR 3 | NP_689995.3 | |||
XM_017011930.1 | 1328 | UTR 3 | XP_016867419.1 |