Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 615137 | |||||||||||||||||||||||||||||||||||||||||
Literature Links: |
WBSCR17 PubMed Links | |||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
WBSCR17 - Williams-Beuren syndrome chromosome region 17 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_022479.2 | 276 | UTR 5 | NP_071924.1 | |||
XM_011516467.2 | 276 | UTR 5 | XP_011514769.1 | |||
XM_011516469.2 | 276 | UTR 5 | XP_011514771.1 | |||
XM_017012521.1 | 276 | UTR 5 | XP_016868010.1 |
Set Membership: |
HapMap |