Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTTTGGAAAAATTGGCATTTTCGGT[C/T]AGTACCCAGTTTTGTAATTTTTTAG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 613036 | ||||||||||||||||||||
Literature Links: |
PMPCA PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
PMPCA - peptidase, mitochondrial processing alpha subunit | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001282944.1 | Intron | NP_001269873.1 | ||||
NM_001282946.1 | Intron | NP_001269875.1 | ||||
NM_015160.2 | Intron | NP_055975.1 | ||||
XM_005266059.4 | Intron | XP_005266116.1 | ||||
XM_011518417.2 | Intron | XP_011516719.1 | ||||
XM_017014543.1 | Intron | XP_016870032.1 |
SDCCAG3 - serologically defined colon cancer antigen 3 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |