Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGACGCCTTCTCCACGCGCTATACT[C/T]ATACGGACCAATGGGTGCGCCCTGT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 614394 MIM: 612912 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
IFT20 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
IFT20 - intraflagellar transport 20 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
TMEM97 - transmembrane protein 97 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_014573.2 | 6 | Intron | NP_055388.2 | |||
XM_005257965.3 | 6 | UTR 5 | XP_005258022.1 |