Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCTGAGCCAACAGGAGCGTCTGGGG[A/G]CAGACATTGATGGTGTACATCCAAG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 610545 MIM: 610546 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
IGFL2 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
IGFL2 - IGF like family member 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001002915.2 | 2636 | Intron | NP_001002915.2 | |||
NM_001135113.1 | 2636 | Intron | NP_001128585.1 | |||
XM_011526501.1 | 2636 | Intron | XP_011524803.1 | |||
XM_017026335.1 | 2636 | Intron | XP_016881824.1 |
IGFL3 - IGF like family member 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_207393.1 | 2636 | Intron | NP_997276.1 | |||
XM_011526953.2 | 2636 | Intron | XP_011525255.1 | |||
XM_017026796.1 | 2636 | Missense Mutation | XP_016882285.1 | |||
XM_017026797.1 | 2636 | Missense Mutation | XP_016882286.1 |