Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTCAGGTTTGGCTGGCTGGGGCGCC[G/T]CCCCCCACCCGAGTACCTTGGGTGG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 611803 MIM: 172490 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
ITFG1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
ITFG1 - integrin alpha FG-GAP repeat containing 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001305002.1 | Intron | NP_001291931.1 | ||||
NM_030790.4 | Intron | NP_110417.2 |
PHKB - phosphorylase kinase regulatory subunit beta | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_000293.2 | Intron | NP_000284.1 | ||||
NM_001031835.2 | Intron | NP_001027005.1 | ||||
XM_005255983.4 | Intron | XP_005256040.1 | ||||
XM_005255984.4 | Intron | XP_005256041.1 | ||||
XM_017023282.1 | Intron | XP_016878771.1 | ||||
XM_017023283.1 | Intron | XP_016878772.1 | ||||
XM_017023284.1 | Intron | XP_016878773.1 |