Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CATCCCTAAACCTGCCCCGGCTTCC[C/T]CTCCTTCCCCCATCCTAGGGGCCAG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 606374 MIM: 104160 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
B3GAT3 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
B3GAT3 - beta-1,3-glucuronyltransferase 3 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
GANAB - glucosidase II alpha subunit | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001278192.1 | Intron | NP_001265121.1 | ||||
NM_001278193.1 | Intron | NP_001265122.1 | ||||
NM_001278194.1 | Intron | NP_001265123.1 | ||||
NM_198334.2 | Intron | NP_938148.1 | ||||
NM_198335.3 | Intron | NP_938149.2 | ||||
XM_017017412.1 | Intron | XP_016872901.1 | ||||
XM_017017413.1 | Intron | XP_016872902.1 | ||||
XM_017017414.1 | Intron | XP_016872903.1 |