Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ATAGGAACACGTATCAAATACCCAG[A/T]AACTTAAGTTATACTCTCTAACAAT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 603128 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
SIM1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
SIM1 - single-minded family bHLH transcription factor 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_005068.2 | 3197 | UTR 3 | NP_005059.2 | |||
XM_005267100.2 | 3197 | UTR 3 | XP_005267157.1 | |||
XM_011536072.2 | 3197 | Intron | XP_011534374.1 | |||
XM_017011197.1 | 3197 | UTR 3 | XP_016866686.1 |