Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTGATCCTGGAATAAGAGCCTGTCC[C/T]GTTAATTTTGGATCTGAAGAATGGA
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 611766 | |||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
MTFMT PubMed Links | |||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | ||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
MTFMT - mitochondrial methionyl-tRNA formyltransferase | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_139242.3 | 1074 | Silent Mutation | ACA,ACG | T,T 302 | NP_640335.2 | |
XM_005254158.4 | 1074 | Silent Mutation | ACA,ACG | T,T 353 | XP_005254215.2 |
Set Membership: |
HapMap |