Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GTAATCACTAGGGATCAAAACTCCT[C/T]AGTCTGGTTGATTGCTGAATGGGAG
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 614660 | |||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
PATL1 PubMed Links | |||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese)
|
||||||
EUR
|
||||||||
AMR
|
PATL1 - PAT1 homolog 1, processing body mRNA decay factor | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_152716.2 | 4762 | UTR 3 | NP_689929.2 | |||
XM_005273833.4 | 4762 | UTR 3 | XP_005273890.1 | |||
XM_005273834.1 | 4762 | UTR 3 | XP_005273891.1 | |||
XM_011544811.2 | 4762 | UTR 3 | XP_011543113.1 | |||
XM_017017339.1 | 4762 | UTR 3 | XP_016872828.1 |
Set Membership: |
HapMap |