Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | |||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||
Phenotype: |
MIM: 614360 MIM: 606079 | |||||||||||||||||||||||||||||
Literature Links: |
ACSM4 PubMed Links | |||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU)
|
|||
EAS - Not Available | African American - Not Available | YRI (Yoruba)
|
|||
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR - Not Available | Japanese - Not Available | JPT (Japanese)
|
|||
EUR - Not Available | |||||
AMR - Not Available |
ACSM4 - acyl-CoA synthetase medium-chain family member 4 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001080454.1 | 1069 | Nonsense Mutation | CAG,TAG | Q,* 357 | NP_001073923.1 |
CD163L1 - CD163 molecule like 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001297650.1 | 1069 | Intron | NP_001284579.1 | |||
NM_174941.5 | 1069 | Intron | NP_777601.2 | |||
XM_011520616.1 | 1069 | Intron | XP_011518918.1 | |||
XM_011520617.2 | 1069 | Intron | XP_011518919.1 | |||
XM_011520618.2 | 1069 | Intron | XP_011518920.1 | |||
XM_011520619.1 | 1069 | Intron | XP_011518921.1 | |||
XM_011520620.1 | 1069 | Intron | XP_011518922.1 | |||
XM_011520621.2 | 1069 | Intron | XP_011518923.1 |
Set Membership: |
HapMap |