Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GCTTTTTGATAGCTTCAATCCACTC[A/G]GCTCGCTCAGCCTTGCTGCTGGCCT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 603907 MIM: 603930 MIM: 608007 | ||||||||||||||||||||
Literature Links: |
EIF2S1 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
EIF2S1 - eukaryotic translation initiation factor 2 subunit alpha | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_004094.4 | 1042 | Intron | NP_004085.1 |
PLEK2 - pleckstrin 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_016445.2 | 1042 | Silent Mutation | GCC,GCT | A,A 343 | NP_057529.1 | |
XM_011536644.2 | 1042 | Silent Mutation | GCC,GCT | A,A 309 | XP_011534946.1 |