Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AACTGGCAGCTGGGGCGTGGGGGTC[A/G]GTTGTCTGGCTGGGCTGTCTCTTCT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 124089 MIM: 611557 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
COX6B1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
COX6B1 - cytochrome c oxidase subunit 6B1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
UPK1A - uroplakin 1A | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001281443.1 | Intron | NP_001268372.1 | ||||
NM_007000.3 | Intron | NP_008931.1 | ||||
XM_017026225.1 | Intron | XP_016881714.1 |
UPK1A-AS1 - UPK1A antisense RNA 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |