Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGGCACTGAGTGCTTCGCAGCTGTC[C/T]GGGCGAGAGGCACAGCGATGGGCTC
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 610432 | |||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
RNF125 PubMed Links | |||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese)
|
||||||
EUR
|
||||||||
AMR
|
RNF125 - ring finger protein 125 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_017831.3 | 634 | UTR 5 | NP_060301.2 | |||
XM_011526045.2 | 634 | UTR 5 | XP_011524347.1 | |||
XM_011526046.2 | 634 | UTR 5 | XP_011524348.1 | |||
XM_011526047.2 | 634 | UTR 5 | XP_011524349.1 |