Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TCTATTGTTTATTTTTTTCATAGAA[C/T]AAAATGTGTTTCTTAGCATATTTTA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 615904 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
PCMTD1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
PCMTD1 - protein-L-isoaspartate (D-aspartate) O-methyltransferase domain containing 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001286782.1 | Intron | NP_001273711.1 | ||||
NM_001286783.1 | Intron | NP_001273712.1 | ||||
NM_052937.3 | Intron | NP_443169.2 | ||||
XM_005251146.2 | Intron | XP_005251203.1 | ||||
XM_011517444.2 | Intron | XP_011515746.1 | ||||
XM_017013017.1 | Intron | XP_016868506.1 |
PXDNL - peroxidasin like | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |