Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGCTGGGTCTAGGTACCCCTGGGGA[C/G]AGGAAGCCTTCGACAAGGCCAGGAA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 607264 MIM: 613939 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
CACNA1G-AS1 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
CACNA1G-AS1 - CACNA1G antisense RNA 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
EPN3 - epsin 3 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
LOC105371824 - uncharacterized LOC105371824 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
SPATA20 - spermatogenesis associated 20 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001258372.1 | 382 | Missense Mutation | CAG,GAG | Q,E 88 | NP_001245301.1 | |
NM_001258373.1 | 382 | Missense Mutation | CAG,GAG | Q,E 44 | NP_001245302.1 | |
NM_022827.3 | 382 | Missense Mutation | CAG,GAG | Q,E 104 | NP_073738.2 |