Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGGTTGGGACGTTGAAATGAGCGCG[C/T]GAGTGGTACGTCCTCTCTCCGCGCT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 608202 | ||||||||||||||||||||
Literature Links: |
CDK5RAP3 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
CDK5RAP3 - CDK5 regulatory subunit associated protein 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001278197.1 | Intron | NP_001265126.1 | ||||
NM_001278198.1 | Intron | NP_001265127.1 | ||||
NM_001278216.1 | Intron | NP_001265145.1 | ||||
NM_001278217.1 | Intron | NP_001265146.1 | ||||
NM_176096.2 | Intron | NP_788276.1 |